Anti-REST Rabbit Polyclonal Antibody
BIRBORB43409-100
: Biorbyt
- Antibody type:Primary
- Antigen name:RE1-silencing transcription factor
- Antigen symbol:REST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: REST (RE1-silencing transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human