Order Entry
Italy
ContactUsLinkComponent
 
Anti-PDCD1LG2 Rabbit Monoclonal Antibody
  BIRBORB42959-100
 :  Biorbyt
Anti-PDCD1LG2 Rabbit Monoclonal Antibody
  BIRBORB42959-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Programmed cell death 1 ligand 2
  • Antigen symbol:
    PDCD1LG2
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human PD-L2 protein.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: PDCD1LG2 (programmed cell death 1 ligand 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human