Order Entry
Italy
ContactUsLinkComponent
Anti-interleukin 17 receptor C isoform 1 precursor Rabbit Polyclonal Antibody
Anti-interleukin 17 receptor C isoform 1 precursor Rabbit Polyclonal Antibody
Catalog # BIRBORB41158-100
Supplier:  Biorbyt
Anti-interleukin 17 receptor C isoform 1 precursor Rabbit Polyclonal Antibody
Catalog # BIRBORB41158-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    interleukin 17 receptor C isoform 1 precursor
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    62 to 471
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 62 and 471 of Human IL17RC.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Western Blot: 1:500-1:3000

Type: Primary
Antigen: interleukin 17 receptor C isoform 1 precursor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human