Anti-HTR2A Rabbit Polyclonal Antibody
100 µg
BIRBORB27585-100
Marketsource
: Biorbyt
Type: Primary
Antigen: HTR2A (5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Form:lyophilized
- Amino acid number:418 to 432
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Sequence:AYKSSQLQMGQKKNS
- Concentration:500 ug/ml
- Shipping temperature:Ice
- Antigen name:5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled
- Cat. No.:BIRBORB27585-100
- Reactivity:Human|Rat|Mouse
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human HTR2A (418-432aa AYKSSQLQMGQKKNS ), different from the mouse sequence by one amino acid.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:HTR2A
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



