Order Entry
Italy
ContactUsLinkComponent
Anti-Lung carcinoma Cluster 2 (MOC-31) Mouse Monoclonal Antibody
Anti-Lung carcinoma Cluster 2 (MOC-31) Mouse Monoclonal Antibody
Catalog # BIRBORB27424-3
Supplier:  Biorbyt
Anti-Lung carcinoma Cluster 2 (MOC-31) Mouse Monoclonal Antibody
Catalog # BIRBORB27424-3
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lung carcinoma Cluster 2
  • Antigen symbol:
    MOC-31
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Concentration:
    neat
  • Shipping temperature:
    Ice
  • Immunogen:
    Preparation of a SCLC-derived cell line
  • Size:
    3 ml
  • Pk:
    3 mL

Specifications

About this item

Type: Primary
Antigen: Lung carcinoma Cluster 2 (MOC-31)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human