Order Entry
Italy
ContactUsLinkComponent
Anti-3-DG-Imidazolon (Advanced Glycation End Product) Mouse Monoclonal Antibody
Anti-3-DG-Imidazolon (Advanced Glycation End Product) Mouse Monoclonal Antibody
Catalog # BIRBORB27342-50
Supplier:  Biorbyt
Anti-3-DG-Imidazolon (Advanced Glycation End Product) Mouse Monoclonal Antibody
Catalog # BIRBORB27342-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3-DG-Imidazolon
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Format:
    concentrated
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    3-DG-derived Imidazolone-HSA
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Immunochemistry: 7 ?g/ml

Type: Primary
Antigen: 3-DG-Imidazolon (Advanced Glycation End Product)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: