Anti-Ricin, RCA60 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB24106-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:Ricin, RCA60
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1
- Storage buffer:PBS, pH 7,4 containing 1% BSA and 50% glycerol
- Storage temperature:Cool
- Concentration:10,0 mg/ml (OD280, E0,1%=1,4)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Ricin, RCA60
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: