Order Entry
Italy
ContactUsLinkComponent
Anti-Group A Strep. Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-Group A Strep. Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB23193-1
Supplier:  Biorbyt
Anti-Group A Strep. Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB23193-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Group A Strep.
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Storage buffer:
    PBS containing 10mg/ml BSA
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Streptococci, group A
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: Group A Strep.
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: