Anti-E. coli all ag's Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB23071-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:E. coli all ag's
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Storage buffer:PBS containing 10mg/ml BSA
- Storage temperature:Cool
- Concentration:1-2 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Mixture of E.coli Serotypes
- Purification:purified by Protein A chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: E. coli all ag's
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: