Order Entry
Italy
ContactUsLinkComponent
Anti-CBR3 Mouse Monoclonal Antibody
Anti-CBR3 Mouse Monoclonal Antibody
Supplier:  Biorbyt
Certificates
Anti-CBR3 Mouse Monoclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB22759-5
  • BIRBORB22759-20
  • BIRBORB22759-100
  • Antigen symbol:
  • CBR3
  • CBR3
  • CBR3
  • Antigen name:
  • carbonyl reductase 3
  • carbonyl reductase 3
  • carbonyl reductase 3
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Anti-human CBR3 mAb is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human CBR3 amino acids 1-277 purified from E.coli
  • Anti-human CBR3 mAb is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human CBR3 amino acids 1-277 purified from E.coli
  • Anti-human CBR3 mAb is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human CBR3 amino acids 1-277 purified from E.coli
  • Amino acid number:
  • 1 to 277
  • 1 to 277
  • 1 to 277
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together