Anti-IgA/IgG/IgM Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB22073-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgA/IgG/IgM
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,6, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified polyclonal human IgG, and IgA and IgM containing factions isolated from human serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: IgG/IgA/IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype: none
Reactivity: