Anti-Rat AlbuminAlbumin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB21672-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:Rat Albumin
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Enzyme/IgG protein molar ratio (E/P) is approximately 1,7, no foreign proteins added,
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: Rat Albumin
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: