Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody
BIRBORB21540-10
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG, including all known subclasses, isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:10 mg
- Pk:10 mg
Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: