Anti-IgM Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
BIRBORB21529-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Rabbit
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 2,7, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified polyclonal IgM and pools of homogenous IgM isolated from pooled human serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: