Anti-Lactalbumin alphaalpha Lactalbumin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21518-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:Lactalbumin alpha
- Antigen symbol:LALBA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,4, no foreign proteins added,
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: alpha Lactalbumin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: