Anti-Mouse Serum Proteins Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21457-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:Mouse Serum Proteins
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,5, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Pooled whole serum and selected protein fractions isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: Mouse Serum Proteins
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: