Anti-IgG1/IgG2a/IgG2b/IgG3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
BIRBORB21451-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG3
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Goat
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Concentration:6,7 mg/ml, Enzyme/IgG protein molar ratio (E/P) is approximately 1,7, No foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG including all known subclasses isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: