Anti-Bence Jones Kappa Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB21180-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Bence Jones Kappa
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Reactivity:Human
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:0,4 mg/ml, Peroxidase/IgG protein molar ratio (E/P) approximately 1,7, No foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Highly purified Bence Jones kappa proteins isolated from pooled human urine.
- Size:200 μg
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: Bence Jones Kappa
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human