Order Entry
Italy
ContactUsLinkComponent
Anti-Protocadherin 16 Rabbit Polyclonal Antibody
Anti-Protocadherin 16 Rabbit Polyclonal Antibody
Catalog # BIRBORB13163-100
Supplier:  Biorbyt
undefined
Anti-Protocadherin 16 Rabbit Polyclonal Antibody
Catalog # BIRBORB13163-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Protocadherin 16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This antibody is produced by immunizing rabbits with a synthetic peptide (KLH-coupled) corresponding to Protocadherin-16.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Western Blot: 1500-1000; Immunofluorescence: 150-100

Type: Primary
Antigen: Protocadherin 16
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human