Order Entry
Italy
ContactUsLinkComponent
Anti-Melamine (1B12) Mouse monoclonal Antibody [clone: 1B12] HRP (Horseradish Peroxidase)
Anti-Melamine (1B12) Mouse monoclonal Antibody [clone: 1B12] HRP (Horseradish Peroxidase)
Catalog # BIRBORB4222-100
Supplier:  Biorbyt
Anti-Melamine (1B12) Mouse monoclonal Antibody [clone: 1B12] HRP (Horseradish Peroxidase)
Catalog # BIRBORB4222-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melamine
  • Clonality:
    Monoclonal
  • Clone:
    1B12
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    0.126 kDa
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    KLH conjugated Melamine.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Melamine (1B12)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: