Order Entry
Italy
ContactUsLinkComponent
 
Anti-Hepatitis C Virus NS4B Rabbit Polyclonal Antibody (FITC (Fluorescein))
  BIRBORB3326-100
 :  Biorbyt
Anti-Hepatitis C Virus NS4B Rabbit Polyclonal Antibody (FITC (Fluorescein))
  BIRBORB3326-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Hepatitis C Virus NS4B
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: Hepatitis C Virus NS4B
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus