Anti-POLR1B Mouse Polyclonal Antibody
ABNOH00084172-A01
: Abnova
- Antibody type:Primary
- Antigen name:Polymerase (RNA) I polypeptide B, 128kDa
- Antigen symbol:POLR1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:84172
- Antigen synonyms:polymerase (RNA) I subunit B|RPA135|RPA2|Rpo1-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: POLR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human