Anti-PLA2G12A Mouse Monoclonal Antibody [clone: 1D11]
ABNOH00081579-M01
: Abnova
- Antibody type:Primary
- Antigen name:Phospholipase A2 Group XIIA
- Antigen symbol:PLA2G12A
- Clonality:Monoclonal
- Clone:1D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:81579
- Antigen synonyms:ROSSY|GXII|PLA2G12
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PLA2G12A
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human