Anti-APOL6 Mouse Monoclonal Antibody [clone: 1F7]
Catalog # ABNOH00080830-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein L6
- Antigen symbol:APOL6
- Clonality:Monoclonal
- Clone:1F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80830
- Antigen synonyms:FLJ38562|DKFZp667M075|APOL-VI|Apolipoprotein L|6|FLJ90164|MGC57495|Apolipoprotein L-VI|Apolipoprotein L6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: APOL6
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human