Anti-ZNF34 Mouse Monoclonal Antibody [clone: 1B11]
ABNOH00080778-M03
: Abnova
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 34
- Antigen symbol:ZNF34
- Clonality:Monoclonal
- Clone:1B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80778
- Antigen synonyms:KOX32
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZNF34
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human