Anti-ADAM33 Mouse Polyclonal Antibody
Catalog # ABNOH00080332-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 33
- Antigen symbol:ADAM33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:80332
- Antigen synonyms:disintegrin and reprolysin metalloproteinase family protein|MGC71889|a disintegrin and metalloprotease 33|DJ964F7.1|FLJ35308|a disintegrin and metalloproteinase domain 33|MGC149823|C20orf153|DKFZp434K0521|RP5-964F7.2|ADAM33|HGNC:15478|FLJ36751|metalloprotease disintegrin|ADAM metallopeptidase domain 33
- Storage buffer:50% glycerol
- Sequence:AHGNCGQDSEGHFLPCAGRDALCGKLQCQGGKPSLLAPHMVPVDSTVHLDGQEVTCRGALALPSAQLDLLGLGLVEPGTQCGPRMVCQSRRCRKNAFQEL
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:ADAM33 (NP_079496, 551 a.a. ~ 650 a.a) partial recombinant protein with GST tag
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Mouse polyclonal antibody raised against a partial recombinant ADAM33.
Recommended Dilutons: Western Blot (1:500 to 1:1000).
Type: Primary
Antigen: ADAM33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human