Anti-ABTB1 Mouse Monoclonal Antibody [clone: 2A10]
Catalog # ABNOH00080325-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ankyrin Repeat And BTB (POZ) Domain Containing 1
- Antigen symbol:ABTB1
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:80325
- Antigen synonyms:BPOZ|Btb3|EF1ABP|PP2259|MGC20585|elongation factor 1A binding protein|ABTB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ABTB1
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human