Anti-GEMIN7 Rabbit Polyclonal Antibody
ABNOH00079760-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Gem (nuclear organelle) associated protein 7
- Antigen symbol:GEMIN7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79760
- Antigen synonyms:Gemin7|SIP3|Gemin-7|GEMI7_HUMAN.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GEMIN7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse