Order Entry
Italy
ContactUsLinkComponent
Anti-PYCRL Mouse Monoclonal Antibody [clone: 4F11]
Anti-PYCRL Mouse Monoclonal Antibody [clone: 4F11]
  ABNOH00065263-M01
 :  Abnova
undefined
Anti-PYCRL Mouse Monoclonal Antibody [clone: 4F11]
  ABNOH00065263-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Pyrroline-5-carboxylate reductase-like
  • Antigen symbol:
    PYCRL
  • Clonality:
    Monoclonal
  • Clone:
    4F11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    65263
  • Antigen synonyms:
    PYCR3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: PYCRL
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human