Anti-RGS18 Mouse Monoclonal Antibody [clone: 1H6]
Catalog # ABNOH00064407-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Regulator of G-protein signaling 18
- Antigen symbol:RGS18
- Clonality:Monoclonal
- Clone:1H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:64407
- Antigen synonyms:RGS13|RP11-142L4.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: RGS18
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human