Anti-RNF25 Mouse Monoclonal Antibody [clone: 1D10]
Catalog # ABNOH00064320-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 25
- Antigen symbol:RNF25
- Clonality:Monoclonal
- Clone:1D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:64320
- Antigen synonyms:AO7|E3 ubiquitin-protein ligase RNF25|RNF25_HUMAN.|RNF 25|E3 ubiquitin protein ligase RNF25|RING finger protein AO7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RNF25
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human