Anti-EDA2R Rabbit Polyclonal Antibody
Catalog # ABNOH00060401-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ectodysplasin A2 Receptor
- Antigen symbol:EDA2R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:60401
- Antigen synonyms:X-linked ectodysplasin-A2 receptor|UNQ2448/PRO5727/PRO34080|XEDAR|Tumor necrosis factor receptor superfamily member 27|EDA-A2 receptor|EDA-A2R|TNFRSF27|EDAA2R
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EDA2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse