Anti-AGGF1 Mouse Polyclonal Antibody
Catalog # ABNOH00055109-B02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Angiogenic Factor With G Patch And FHA Domains 1
- Antigen symbol:AGGF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:55109
- Antigen synonyms:HSU84971|GPATCH7|HUS84971|VG5Q|GPATC7
- Storage buffer:No additive
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:AGGF1 (AAH32844.1, 1 a.a. ~ 714 a.a) full-length human protein
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human AGGF1 protein.
Recommended Dilutons: Western Blot (1:500 to 1:1000).
Type: Primary
Antigen: AGGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human