Anti-SETD4 Mouse Polyclonal Antibody
50 µl
Catalog # ABNOH00054093-B01
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: SETD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00054093-B01.pdf
Datasheet
Specifications
- Size:50 µl
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:SET Domain Containing 4
- Cat. No.:ABNOH00054093-B01
- Reactivity:Human
- Gene ID:54093
- Antigen synonyms:C21orf18|C21orf27
- Conjugation:Unconjugated
- Antigen symbol:SETD4
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



