Anti-NOP16 Mouse Monoclonal Antibody [clone: 1G4]
Catalog # ABNOH00051491-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nucleolar protein 16
- Antigen symbol:NOP16
- Clonality:Monoclonal
- Clone:1G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51491
- Antigen synonyms:HSPC185|HSPC111
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NOP16
Clonality: Monoclonal
Clone: 1G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human