Anti-FLVCR1 Mouse Monoclonal Antibody [clone: 4B2]
Catalog # ABNOH00028982-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Feline leukemia virus subgroup C cellular receptor 1
- Antigen symbol:FLVCR1
- Clonality:Monoclonal
- Clone:4B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- ChIP:Yes
- Gene ID:28982
- Antigen synonyms:eline leukemia virus subgroup C cellular receptor 1|AXPC1|feline leukemia virus subgroup C cellular receptor|FLVCR|PCA|FLVCR1|FLVCR protein|MFSD7B|PCARP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FLVCR1
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human