Anti-ADAMDEC1 Mouse Monoclonal Antibody [clone: 6C4]
Catalog # ABNOH00027299-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ADAM-like, Decysin 1
- Antigen symbol:ADAMDEC1
- Clonality:Monoclonal
- Clone:6C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:27299
- Antigen synonyms:M12.219
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ADAMDEC1
Clonality: Monoclonal
Clone: 6C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human