Anti-HAVCR1 Mouse Monoclonal Antibody [clone: 2B9]
Catalog # ABNOH00026762-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:hepatitis A virus cellular receptor 1
- Antigen symbol:HAVCR1
- Clonality:Monoclonal
- Clone:2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:26762
- Antigen synonyms:TIM1|TIM-1|KIM-1|CD365|HAVCR|HAVCR-1|TIMD1|TIMD-1|TIM|KIM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HAVCR1
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human