Anti-PTPN22 Mouse Monoclonal Antibody [clone: 4F6]
Catalog # ABNOH00026191-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase, non-receptor type 22
- Antigen symbol:PTPN22
- Clonality:Monoclonal
- Clone:4F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:26191
- Antigen synonyms:PEP|PTPN22|PTPN8|PEST-domain phosphatase|LyP|Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP|Lymphoid phosphatase|Tyrosine-protein phosphatase non-receptor type 22
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PTPN22
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human