Anti-LDOC1 Mouse Polyclonal Antibody
ABNOH00023641-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Leucine zipper protein down-regulated in cancer cells
- Antigen symbol:LDOC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23641
- Antigen synonyms:Leucine zipper protein down-regulated in cancer cells|Protein LDOC1|LDOC1|Mar7|BCUR1|Mart7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: LDOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human