Anti-CLCF1 Mouse Monoclonal Antibody [clone: 7E10]
ABNOH00023529-M01A
: Abnova
- Antibody type:Primary
- Antigen name:Cardiotrophin-like cytokine factor 1
- Antigen symbol:CLCF1
- Clonality:Monoclonal
- Clone:7E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23529
- Antigen synonyms:CISS2|BSF-3|NR6|BSF3|CLC|NNT-1|NNT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: CLCF1
Clonality: Monoclonal
Clone: 7E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human