Anti-TNRC6B Mouse Monoclonal Antibody [clone: 4B11]
ABNOH00023112-M02
: Abnova
- Antibody type:Primary
- Antigen name:Trinucleotide Repeat Containing 6B
- Antigen symbol:TNRC6B
- Clonality:Monoclonal
- Clone:4B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23112
- Antigen synonyms:TNR6B_HUMAN.|TNRC 6B|Trinucleotide repeat containing 6B|Trinucleotide repeat containing gene 6B protein|KIAA1093
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TNRC6B
Clonality: Monoclonal
Clone: 4B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human