Anti-ADGRL1 Mouse Monoclonal Antibody [clone: 4E12]
Catalog # ABNOH00022859-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adhesion G Protein-Coupled Receptor L1
- Antigen symbol:ADGRL1
- Clonality:Monoclonal
- Clone:4E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:22859
- Antigen synonyms:CL1|LEC2|LPHN1|CIRL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ADGRL1
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human