Anti-CA5B Mouse Polyclonal Antibody
Catalog # ABNOH00011238-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 5B, mitochondrial
- Antigen symbol:CA5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11238
- Antigen synonyms:CA-VB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CA5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human