Anti-PMAIP1 Rabbit Polyclonal Antibody
Catalog # ABNOH00010811-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phorbol-12-myristate-13-acetate-induced protein 1
- Antigen symbol:PMAIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10811
- Antigen synonyms:Immediate-early-response protein APR|PMA-induced protein 1|NOXA|Protein Noxa|Phorbol-12-myristate-13-acetate-induced protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PMAIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human