Order Entry
Italy
ContactUsLinkComponent
 
Anti-ZNF274 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # ABNOH00010782-M02
Supplier:  Abnova
undefined
Anti-ZNF274 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # ABNOH00010782-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 274
  • Antigen symbol:
    ZNF274
  • Clonality:
    Monoclonal
  • Clone:
    2F5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    10782
  • Antigen synonyms:
    HFB101|ZF2|ZKSCAN19|ZSCAN51
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: ZNF274
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human