Anti-MGEA5 Mouse Monoclonal Antibody [clone: 1C7]
ABNOH00010724-M02
: Abnova
- Antibody type:Primary
- Antigen name:Meningioma expressed antigen 5 (hyaluronidase)
- Antigen symbol:MGEA5
- Clonality:Monoclonal
- Clone:1C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10724
- Antigen synonyms:MEA5|NCOAT|OGA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MGEA5
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human