Anti-CD96 Mouse Monoclonal Antibody [clone: 1C8]
ABNOH00010225-M02
: Abnova
- Antibody type:Primary
- Antigen name:CD96 molecule
- Antigen symbol:CD96
- Clonality:Monoclonal
- Clone:1C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10225
- Antigen synonyms:Increased Late Expression|Tactile|1700109I12Rik|T Cell Activation|T Cell-Activated Increased Late Expression Protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CD96
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human