Order Entry
Italy
ContactUsLinkComponent
 
Anti-DHRS9 Rabbit Polyclonal Antibody
  ABNOH00010170-D01P
 :  Abnova
undefined
Anti-DHRS9 Rabbit Polyclonal Antibody
  ABNOH00010170-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Dehydrogenase/reductase (SDR family) Member 9
  • Antigen symbol:
    DHRS9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10170
  • Antigen synonyms:
    SDR9C4|RDHTBE|RDHL|RDH15|RDH-E2|RDH-TBE|3ALPHA-HSD|RETSDR8|3-alpha-HSD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: DHRS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human