Anti-DNAJB6 Mouse Polyclonal Antibody
Catalog # ABNOH00010049-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DnaJ (Hsp40) homolog, subfamily B, member 6
- Antigen symbol:DNAJB6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10049
- Antigen synonyms:DnaJ|HSJ-2|MSJ-1|HHDJ1|MRJ|LGMD1E|HSJ2|LGMD1D|DJ4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DNAJB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human